Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028006.2 | NP_082282.1 | 9 | 887 | |
GenBank mRNA | AK010005.1 | - | 9 | 881 | |
AK040492.1 | - | 1 | 2445 | ||
AK041416.1 | - | 9 | 852 | ||
AK041986.1 | - | 293 | |||
AK087403.1 | - | 8 | 666 | ||
AK148039.1 | - | 9 | 887 | ||
BC038654.1 | - | 6 | 852 | ||
BC062179.1 | - | 9 | 884 | 6333817 |