Gene Transcripts

Gene Symbol
Fundc1
Entrez Gene ID
72018
Gene Name
FUN14 domain containing 1
Gene Aliases
1500005J14Rik, 1810033P05Rik
Chromosome Location
Chr. X: 17519967 - 17572330 on Build GRCm38
UniGene ID
Mm.41558
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028058.3 NP_082334.1 2, 3 301
GenBank mRNA AK005163.1 - 2, 3 302
AK007686.1 - 1, 2 338
AK168868.1 - 2, 3 288
AK210161.1 - 1, 2 62
BC012515.1 - 2, 3 243 2651125
BC030487.1 - 2, 3 307 5374103

View Full Product Details