Gene Transcripts

Gene Symbol
Fbxo47
Entrez Gene ID
72973
Gene Name
F-box protein 47
Gene Aliases
2900052P03Rik, Gm1562
Chromosome Location
Chr. 11: 97851558 - 97884390 on Build GRCm38
UniGene ID
Mm.245939
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081435.1 NP_001074904.1 10 1388
GenBank mRNA AK013672.1 - 1 90
BC099560.1 - 1 84
BC158049.1 - 10 1323 9007353

View Full Product Details