Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033371.2 | NP_203535.1 | 2 | 894 | |
GenBank mRNA | AK154865.1 | - | 2 | 879 | |
AK171452.1 | - | 2 | 879 | ||
AK220248.1 | - | 2 | 894 | ||
AY010723.1 | - | 1 | 89 | ||
BC033318.1 | - | 1 | 454 | ||
BC044775.1 | - | 1 | 576 | ||
BC049958.1 | - | 1 | 576 | ||
BC057450.1 | - | 1 | 511 | 3966334 | |
BC096620.1 | - | 2 | 894 | 6390076 |