Gene Transcripts
- Gene Symbol
- Krt80
- Entrez Gene ID
- 74127
- Gene Name
- keratin 80
- Gene Aliases
- 1200016G03Rik, 2310041I20Rik, Kb20
- Chromosome Location
- Chr. 15: 101348315 - 101370125 on Build GRCm38
- UniGene ID
- Mm.440055
- Species
- Mus musculus
- Gene Symbol
- Gm35853
- Entrez Gene ID
- 102639573
- Gene Name
- predicted gene, 35853
- Gene Aliases
- Chromosome Location
- Chr. 15: 101358708 - 101405857 on Build GRCm38
- UniGene ID
- Mm.384081
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138921.1 |
- |
|
545 |
|
View Full Product Details