Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024622.2 | NP_001019793.1 | 2 | 157 | |
GenBank mRNA | AK145195.1 | - | 2 | 116 | |
AK150595.1 | - | 2 | 118 | ||
AK152279.1 | - | 2 | 120 | ||
AK159201.1 | - | 2 | 128 | ||
AK166071.1 | - | 2 | 141 | ||
AK172325.1 | - | 1 | 162 | ||
BC023320.1 | - | 2 | 128 | ||
BC061235.1 | - | 2 | 121 | 6593866 | |
BC132588.1 | - | 2 | 179 | 40130865 | |
BC132590.1 | - | 2 | 179 | 40130867 | |
BY287179.1 | - | 2 | 157 |