Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001200028.1 | NP_001186957.1 | 2, 3 | 371 | |
NM_029604.3 | NP_083880.2 | 2, 3 | 323 | ||
GenBank mRNA | AK006407.1 | - | 2, 3 | 324 | |
AW519725.1 | - | 1, 2 | 35 | ||
BC049757.1 | - | 2, 3 | 372 | 6773491 | |
HM237137.1 | - | 2, 3 | 213 | ||
HM237138.1 | - | 2, 3 | 371 |