Gene Transcripts
- Gene Symbol
- Mzt1
- Entrez Gene ID
- 76789
- Gene Name
- mitotic spindle organizing protein 1
- Gene Aliases
- 2410129H14Rik, AI120158, Mozart1
- Chromosome Location
- Chr. 14: 99034544 - 99046463 on Build GRCm38
- UniGene ID
- Mm.38912
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175245.4 |
NP_780454.1
|
1, 2 |
152 |
|
GenBank mRNA |
AK010788.1 |
- |
1, 2 |
128 |
|
AK082817.1 |
- |
1, 2 |
98 |
|
AK087564.1 |
- |
1, 2 |
96 |
|
AK150268.1 |
- |
1, 2 |
128 |
|
AK151232.1 |
- |
1, 2 |
128 |
|
AK151358.1 |
- |
1, 2 |
128 |
|
AK151819.1 |
- |
1, 2 |
128 |
|
AK160911.1 |
- |
1, 2 |
137 |
|
AK165605.1 |
- |
1, 2 |
115 |
|
AK168114.1 |
- |
1, 2 |
93 |
|
AK169036.1 |
- |
1, 2 |
102 |
|
AK170746.1 |
- |
1, 2 |
102 |
|
BB844588.1 |
- |
1, 2 |
153 |
|
BC038168.1 |
- |
1, 2 |
90 |
|
BC098481.1 |
- |
1, 2 |
115 |
|
BC117005.1 |
- |
1, 2 |
127 |
40126324 |
BC117007.1 |
- |
1, 2 |
127 |
40126326 |
View Full Product Details