Gene Transcripts

Gene Symbol
Spata2l
Entrez Gene ID
78779
Gene Name
spermatogenesis associated 2-like
Gene Aliases
2610039E05Rik, AI648233, mKIAA4138
Chromosome Location
Chr. 8: 123232255 - 123236245 on Build GRCm38
UniGene ID
Mm.41459
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030176.2 NP_084452.2 3 882
GenBank mRNA AK019166.1 - 2 1207
AK081234.1 - 3 882
AK170267.1 - 2 1182
AK220503.1 - 1182
BC139310.1 - 2 817 8862332
BC139311.1 - 2 817 8862333

View Full Product Details