Gene Transcripts
- Gene Symbol
- Tusc2
- Entrez Gene ID
- 80385
- Gene Name
- tumor suppressor candidate 2
- Gene Aliases
- 1190001E22Rik, AA407686, Fus1, Lgcc, PAP, Pdap2
- Chromosome Location
- Chr. 9: 107563255 - 107566108 on Build GRCm38
- UniGene ID
- Mm.21739
- Species
- Mus musculus
- Gene Symbol
- Gm9917
- Entrez Gene ID
- 100038755
- Gene Name
- predicted gene 9917
- Gene Aliases
- ENSMUSG00000053666
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.490992
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK044848.1 |
- |
|
840 |
|
BC094226.1 |
- |
|
489 |
|
View Full Product Details