Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178936.3 | NP_849267.1 | 5 | 548 | |
GenBank mRNA | AK016295.1 | - | 7 | 802 | |
AK147601.1 | - | 5 | 596 | ||
AK149544.1 | - | 5 | 573 | ||
AK163285.1 | - | 5 | 586 | ||
BC040365.1 | - | 5 | 548 | 4223855 |