Product Details

SNP ID
rs6248229
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:60183029 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AATGGTGGAATGCTTGCTTTACGTG[C/T]GCAAGGCCCAGGGTTTGATAACCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Baz2b PubMed Links

Gene Details

Gene
Baz2b
Gene Name
bromodomain adjacent to zinc finger domain, 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001182.3 Intron NP_001001182.2

View Full Product Details