Product Details
- SNP ID
-
rs13476784
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:134519311 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCCTCAGATATAAAGGGACTTCT[A/C]TTTCTATTTTCTTGGGATGACAATG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Hao1
PubMed Links
Gene Details
- Gene
- Hao1
- Gene Name
- hydroxyacid oxidase 1, liver
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010403.2 |
|
Intron |
|
|
NP_034533.1 |
View Full Product Details