Product Details
- SNP ID
-
rs13478762
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:54225677 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACAGAAAAGATTCAAGATCTCAA[A/G]TGGTACCCAAAGGAGGAGGTGGGAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Chn2
PubMed Links
Gene Details
- Gene
- Chn2
- Gene Name
- chimerin 2
View Full Product Details