Product Details

SNP ID
rs13483737
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.X:47876770 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAACAAAATCTCACCTTACATGAAC[C/T]AGAAGAACACTTCATGTCATGAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Smarca1 PubMed Links

Gene Details

Gene
Smarca1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053123.4 Intron NP_444353.3

View Full Product Details