Product Details
- SNP ID
-
rs13483749
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:50932422 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCAAATCTAAGGACCTTATAGTA[A/T]AGCTTTTTAAATAATTCTCAAAGTA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Frmd7
PubMed Links
Gene Details
- Gene
- Frmd7
- Gene Name
- FERM domain containing 7
View Full Product Details