Product Details

SNP ID
rs13481203
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:103732228 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACACTGCATACAGGAAGGCCGTCTC[C/T]TTAAAGCCTGTGCGGGAGAAAAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C130046K22Rik PubMed Links

Gene Details

Gene
C130046K22Rik
Gene Name
RIKEN cDNA C130046K22 gene
There are no transcripts associated with this gene.

Gene
Wnt9b
Gene Name
wingless-type MMTV integration site family, member 9B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011719.4 400 Missense Mutation NP_035849.3

View Full Product Details