Product Details

SNP ID
rs13481366
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:28861603 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCCACATATGTGCACCTGTACTCAC[A/G]TACCTGAATGCACATGTGCCTACAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Tssc1 PubMed Links

Gene Details

Gene
Tssc1
Gene Name
tumor suppressing subtransferable candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201357.2 Intron NP_958745.2

View Full Product Details