Product Details
- SNP ID
-
rs13459086
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:49990206 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGGACAATCCGACTGCAGGCCTT[C/T]GCTTCCCTCTGTGCCAGCCACAGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Adgra2
PubMed Links
Gene Details
- Gene
- Adgra2
- Gene Name
- adhesion G protein-coupled receptor A2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_133911.1 |
1712 |
Silent Mutation |
|
|
NP_598672.1 |
View Full Product Details