Product Details

SNP ID
rs3712347
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:46830176 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AACTTGGTGCAAAGCATGTGCAAGA[T/C]GGCAGTTTTCTACACTGTTTCAGCA
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
Slc39a10 PubMed Links

Gene Details

Gene
Slc39a10
Gene Name
solute carrier family 39 (zinc transporter), member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172653.2 Intron NP_766241.2

View Full Product Details