Product Details
- SNP ID
-
rs13477101
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:49754917 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGATAACTGAGAGTGCCCATTCCG[C/T]AGAGGGAATGGATTCCATGCTGACG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pcdh18
PubMed Links
Gene Details
- Gene
- Pcdh18
- Gene Name
- protocadherin 18
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_130448.3 |
2400 |
Missense Mutation |
|
|
NP_569715.3 |
View Full Product Details