Product Details

SNP ID
rs3697980
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:164012492 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCTAAAACACCTCATGATTATATT[C/T]TTTCCCTCATATCTGGAGCCCACTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ywhab PubMed Links

Gene Details

Gene
Ywhab
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018753.6 Intron NP_061223.2

View Full Product Details