Product Details

SNP ID
rs13483249
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:22379928 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTGGGAAGGTGGCAGACTATTTTA[A/C]TGATGACTTTCTGTATTTGGCTTGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Asxl3 PubMed Links

Gene Details

Gene
Asxl3
Gene Name
additional sex combs like 3 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167777.1 Intron NP_001161249.1

View Full Product Details