Product Details
- SNP ID
-
rs13482862
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:8817709 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCCAGGCCCAACTGAGTACATG[A/G]TGTCTGTACCACGTCTGCCACTGTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pde10a
PubMed Links
Gene Details
- Gene
- Pde10a
- Gene Name
- phosphodiesterase 10A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_011866.2 |
|
Intron |
|
|
NP_035996.2 |
View Full Product Details