Product Details

SNP ID
rs13482849
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:5612314 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGTGTAAATAGCACACGTTTGGGAA[C/T]CTCAACTTCTCATTAAATTATCAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Zdhhc14 PubMed Links

Gene Details

Gene
Zdhhc14
Gene Name
zinc finger, DHHC domain containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_146073.3 Intron NP_666185.3

View Full Product Details