Product Details

SNP ID
rs13476535
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:61756786 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AATTTAACAGAAAAGAGGAATGGCC[A/G]AATGTCACTTTTGTTCTAAAACACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Psmd14 PubMed Links

Gene Details

Gene
Psmd14
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021526.2 Intron NP_067501.2

View Full Product Details