Product Details
- SNP ID
-
rs13480722
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:97649553 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTTTTAAATTACCAGCATACATG[C/T]GTTGTTTTTAAACTATGCTACCAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Epyc
PubMed Links
Gene Details
- Gene
- Epyc
- Gene Name
- epiphycan
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007884.2 |
|
Intron |
|
|
NP_031910.1 |
View Full Product Details