Product Details
- SNP ID
-
rs13483101
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:73340530 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTACAGATCTGTCCAGCTGCTTCCC[A/T]TCCTTTTCATGGACACATGCTAAGT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Capn13
PubMed Links
Gene Details
- Gene
- Capn13
- Gene Name
- calpain 13
View Full Product Details