Product Details
- SNP ID
-
rs6182393
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:28344949 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAAGCCTGTGAAGTTACATGAGGT[C/T]CACTCAGTACAGTCAGCATCTACAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Glis3
PubMed Links
Gene Details
- Gene
- Glis3
- Gene Name
- GLIS family zinc finger 3
There are no transcripts associated with this gene.
View Full Product Details