Product Details

SNP ID
rs13483117
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:78840982 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGGAAAAAGTGGCATAGGTCA[C/T]GAGTCATCATTAAAACGGAAAGCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Gm6548 PubMed Links

Gene Details

Gene
Gm6548
Gene Name
eukaryotic translation elongation factor 1 alpha 1 pseudogene
There are no transcripts associated with this gene.

Gene
Gpatch11
Gene Name
G patch domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181649.6 651 Silent Mutation NP_857632.4
Gene
Heatr5b
Gene Name
HEAT repeat containing 5B
There are no transcripts associated with this gene.

View Full Product Details