Product Details

SNP ID
rs13483156
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:88583915 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGCTGAGCATGATGAGCGACCACCT[A/G]TGCAGCATGAACGAGACCCTGTCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ppp1r21 PubMed Links

Gene Details

Gene
Ppp1r21
Gene Name
protein phosphatase 1, regulatory subunit 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_028658.4 2385 Silent Mutation NP_082934.3

View Full Product Details