Product Details
- SNP ID
-
rs13482479
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:23139632 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTAACTGATTCCTACTACAATGT[A/G]TTGTTAAATTACAGTTCCAACTTGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cdh18
PubMed Links
Gene Details
- Gene
- Cdh18
- Gene Name
- cadherin 18
View Full Product Details