Product Details

SNP ID
rs13477708
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:53144580 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTCTAACATTTCGCTACAGACAAAG[C/T]TGAGCCCTTCAAGTGATAAACGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Abca1 PubMed Links

Gene Details

Gene
Abca1
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013454.3 Intron NP_038482.3

View Full Product Details