Product Details
- SNP ID
-
rs13476906
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:168755443 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGGCCCTGACGTGAGCTTTTGAGG[C/T]AGCCCTAGAGAAGGGGTTCCCGTGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Gm14261
PubMed Links
Gene Details
- Gene
- Gm14261
- Gene Name
- predicted gene 14261
There are no transcripts associated with this gene.
- Gene
- Sall4
- Gene Name
- sal-like 4 (Drosophila)
There are no transcripts associated with this gene.
View Full Product Details