Product Details
- SNP ID
-
rs13481119
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:79547199 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCCGAGCTTCCACACATGGGCTG[C/G]TCATCAATATCATTCACTCTCTGTG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Nf1
PubMed Links
Gene Details
- Gene
- Nf1
- Gene Name
- neurofibromatosis 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010897.2 |
6490 |
Missense Mutation |
|
|
NP_035027.1 |
View Full Product Details