Product Details

SNP ID
rs3090057
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:41737831 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGAAATTCAAGCCCAGTGCCATGGT[G/A]CATGCCGGGAAGCCTCGAACTTGGA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
Oxr1 PubMed Links

Gene Details

Gene
Oxr1
Gene Name
oxidation resistance 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130163.1 Intron NP_001123635.1
NM_001130164.1 Intron NP_001123636.1
NM_001130165.1 Intron NP_001123637.1
NM_001130166.1 Intron NP_001123638.1
NM_130885.2 Intron NP_570955.1

View Full Product Details