Product Details

SNP ID
rs6186902
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.19:26723514 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCCCTATCCCTGGCCTTGATCCTAC[A/G]CCCTGGAAAAGACAGAACAATGCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Smarca2 PubMed Links

Gene Details

Gene
Smarca2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011416.2 Intron NP_035546.2
NM_026003.2 Intron NP_080279.1

View Full Product Details