Product Details

SNP ID
rs3689602
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:30412836 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAGTAGCTCCTTTTCAAAAAT[G/T]TCCATGTAATTCTTCTGCTGAGCAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Crat PubMed Links

Gene Details

Gene
Crat
Gene Name
carnitine acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007760.3 Intron NP_031786.2
Gene
Ppp2r4
Gene Name
protein phosphatase 2A activator, regulatory subunit B
There are no transcripts associated with this gene.

View Full Product Details