Product Details
- SNP ID
-
rs13477674
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:43549182 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCTGCCTGCAGCAGGTTCTGACG[A/G]GGCTGTGGAGAGTGCACACCTGTCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tln1
PubMed Links
Gene Details
- Gene
- Tln1
- Gene Name
- talin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_011602.5 |
2386 |
Silent Mutation |
|
|
NP_035732.2 |
View Full Product Details