Product Details

SNP ID
rs13483389
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:60707868 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CATCAGCACCCAGGTGTGGGATGGA[C/T]TCGGAGGAGCGTGTCTTGGCCAACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ndst1 PubMed Links

Gene Details

Gene
Ndst1
Gene Name
N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008306.4 895 Silent Mutation NP_032332.2

View Full Product Details