Product Details

SNP ID
rs6398113
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:151692393 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTGTAACATTTGGGTCTTCTCACAT[C/T]TATTCACACTCATGCTCAAAATCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Fam129a PubMed Links

Gene Details

Gene
Fam129a
Gene Name
family with sequence similarity 129, member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022018.3 Intron NP_071301.2

View Full Product Details