Product Details
- SNP ID
-
rs13483138
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:84441114 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGCACCTAAAATACATGAAAATAC[A/G]TTAATTTTTCTAAGAAATTAAGAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Thada
PubMed Links
Gene Details
- Gene
- Thada
- Gene Name
- thyroid adenoma associated
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_183021.3 |
|
Intron |
|
|
NP_898842.2 |
View Full Product Details