Product Details
- SNP ID
-
rs13481055
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:60336581 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGCACTGTTTCACTTGGCTGTTC[A/G]TGGGCTGAATCCTTTGCATCAGGGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tom1l2
PubMed Links
Gene Details
- Gene
- Tom1l2
- Gene Name
- target of myb1-like 2 (chicken)
View Full Product Details