Product Details
- SNP ID
-
rs4223708
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:8560978 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACATGTGTTTGGGTTTTAGAAACA[G/A]GTGCATGTGAAGACAAATCTGAGCT
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Stmn2
PubMed Links
Gene Details
- Gene
- Stmn2
- Gene Name
- stathmin-like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025285.2 |
1278 |
UTR 3 |
|
|
NP_079561.1 |
View Full Product Details