Product Details
- SNP ID
-
rs13477404
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:130369997 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCTACTGAAGTCCTCCACAGCGG[A/G]CCTCGGGGAGCCCCCGAAAGCAGCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Col25a1
PubMed Links
Gene Details
- Gene
- Col25a1
- Gene Name
- collagen, type XXV, alpha 1
View Full Product Details