Product Details

SNP ID
rs13480763
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:110943560 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTGCTTCTTGAAAAAAATCCA[C/T]CTTAACTCTTTTAAAAATTGGGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Csrp2 PubMed Links

Gene Details

Gene
Csrp2
Gene Name
cysteine and glycine-rich protein 2
There are no transcripts associated with this gene.

Gene
Zdhhc17
Gene Name
zinc finger, DHHC domain containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172554.2 2768 UTR 3 NP_766142.2

View Full Product Details