Product Details

SNP ID
rs33859939
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:129928766 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTACATAATAAATGTTTATATATA[G/T]AGAGAAGGGGGAGATAGAGAGGGAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Thsd7b PubMed Links

Gene Details

Gene
Thsd7b
Gene Name
thrombospondin, type I, domain containing 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172485.3 Intron NP_766073.2

View Full Product Details