Product Details

SNP ID
rs6299624
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:175643878 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCACAGTCCAGTGGGCCAAGAGACA[C/T]AAGCTGACAGCTTGCCAGGTGCTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Kmo PubMed Links

Gene Details

Gene
Kmo
Gene Name
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133809.1 Intron NP_598570.1

View Full Product Details