Product Details
- SNP ID
-
rs13477643
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:34982748 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACTTAATGTGTGCCTAATCTATT[C/T]AATTTTTATGATTCCATACTCCGTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Mob3b
PubMed Links
Gene Details
- Gene
- Mob3b
- Gene Name
- MOB kinase activator 3B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178061.5 |
|
Intron |
|
|
NP_835162.1 |
View Full Product Details