Product Details

SNP ID
rs3671973
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:32576432 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGCGACACAGTGCATCTCAGTGCT[A/G]AACGAGAGTTTCACTCGTTCATTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Bach2 PubMed Links

Gene Details

Gene
Bach2
Gene Name
BTB and CNC homology 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109661.1 Intron NP_001103131.1
Gene
Bach2os
Gene Name
BTB and CNC homology 2, opposite strand
There are no transcripts associated with this gene.

View Full Product Details